Spermin Tetrahydrochlorid 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine) tetrahydrochloride

BIS5943-10G BIS5943-25G BIS5943-5G
APOSBIS5943-10GEA 168 EUR
APOSBIS5943-10G APOSBIS5943-25G APOSBIS5943-5G
Spermin Tetrahydrochlorid 99%
Spermin Tetrahydrochlorid

For precipitation of DNA from aqueous buffers

Formel: C₁₀H₃₀Cl₄N₄
Schmelzpunkt: 310…311 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00012914
CAS-Nummer: 306-67-2
EINECS: 206-189-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR