3,3',5'-Triiodo-L-thyronine sodium salt

Lieferant: Apollo Scientific
Achtung

Synonyme: Liothyronine sodium salt, Sodium (S)-2-amino-3-(4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl)propanoate

BISC1055-1MG
APOSBISC1055-1MGEA 2820 EUR
APOSBISC1055-1MG
3,3',5'-Triiodo-L-thyronine sodium salt
3,3',5'-Triiodo-L-thyronine sodium salt
Formel: C₁₅H₁₁I₃NNaO₄
Molecular Weight: 672,959 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00002594
CAS-Nummer: 55-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR