Vitamin-D3-[2H3]-Lösung 1 mg/mL in Ethanol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Cholecalciferol
BISC1060A-1ML
APOSBISC1060A-1MLEA
1210
EUR
APOSBISC1060A-1ML
Vitamin-D3-[2H3]-Lösung 1 mg/mL in Ethanol
Vitamin-D3-[2H3]
Formel:
C₂₇H₄₁D₃O Molecular Weight: 387,656 g/mol Dichte: 0,79 g/cm³ (25 °C) Flammpunkt: 14 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD11656127 CAS-Nummer: 80666-48-4 UN: 1170 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...