Desmopressin Acetate 99

Lieferant: Apollo Scientific

BISN0004-5MG BISN0004-25MG BISN0004-50MG
APOSBISN0004-5MGEA 132 EUR
APOSBISN0004-5MG APOSBISN0004-25MG APOSBISN0004-50MG
Desmopressin Acetate 99
Desmopressin Acetate

Desmopressin acetate is the synthetic analogue of the antidiuretic hormone arginine vasopressin.

Formel: C₄₆H₆₄N₁₄O₁₂S₂.₂CH₃CO₂H
Molecular Weight: 1189.321 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00133962
CAS-Nummer: 62288-83-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR