AM-095 freie Säure 98%

Lieferant: Apollo Scientific

BISN0022-1MG BISN0022-5MG BISN0022-10MG
APOSBISN0022-1MGEA 109 EUR
APOSBISN0022-1MG APOSBISN0022-5MG APOSBISN0022-10MG
AM-095 freie Säure 98%
AM-095

AM095 free acid is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 μM for recombinant human or mouse LPA1 respectively.

Formel: C₂₇H₂₄N₂O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD22665734
CAS-Nummer: 1228690-36-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR