AZD-0156 99%
Lieferant: Apollo Scientific
BISN0034-5MG
APOSBISN0034-5MGEA
58
EUR
APOSBISN0034-5MG
AZD-0156 99%
AZD-0156
AZD0156 is a potent and selective inhibitors of ATM kinase, with potential chemo-/radio-sensitizing and antineoplastic activities.
Formel:
C₂₆H₃₁N₅O₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD30470661 CAS-Nummer: 1821428-35-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...