GNE-3511 99%

Lieferant: Apollo Scientific

Synonyme: 2-[[6-(3,3-difluoropyrrolidin-1-yl)-4-[1-(oxetan-3-yl)piperidin-4-yl]pyridin-2-yl]amino]pyridine-4-carbonitrile, 2-((6-(3,3-Difluoropyrrolidin-1-yl)-4-(1-(oxetan-3-yl)piperidin-4-yl)pyridin-2-yl)amino)isonicotinonitrile

BISN0038-1MG
APOSBISN0038-1MGEA 55.1 EUR
APOSBISN0038-1MG
GNE-3511 99%
GNE-3511

GNE-3511 is a potent and selective zipper kinase (DLK, MAP3K12) inhibitors.

Formel: C₂₃H₂₆F₂N₆O
Molecular Weight: 440,49 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28502215
CAS-Nummer: 1496581-76-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR