Edicotinib 99%
Lieferant: Apollo Scientific
Synonyme:
5-Cyano-N-(2-(4,4-dimethylcyclohex-1-en-1-yl)-6-(2,2,6,6-tetramethyltetrahydro-2H-pyran-4-yl)pyridin-3-yl)-1H-imidazole-2-carboxamide, PRV 6527, JNJ 40346527, 4-cyano-N-[2-(4,4-dimethylcyclohex-1-en-1-yl)-6-(2,2,6,6-tetramethyloxan-4-yl)pyridin-3-yl]-1H-imidazole-2-carboxamide
BISN0042-50MG
APOSBISN0042-50MGEA
249
EUR
APOSBISN0042-50MG
Edicotinib 99%
Edicotinib
JNJ-40346527 is an inhibitor of colony-stimulating factor-1 receptor (CSF1R) with potential antineoplastic activity.
Formel:
C₂₇H₃₅N₅O₂ Molecular Weight: 461.61 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30489233 CAS-Nummer: 1142363-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...