SSR-240612 98%

Lieferant: Apollo Scientific

BISN0050-1MG
APOSBISN0050-1MGEA 78.3 EUR
APOSBISN0050-1MG
SSR-240612 98%
SSR-240612

SSR240612 is a potent and selective non-peptide bradykinin B1 receptor antagonist with Kis of 0.48 nM and 0.73 nM for B1 kinin receptors of human fibroblast MRC5 and HEK cells expressing human B1 receptors, 481 nM and 358 nM for B2 receptors of guinea pig ileum membranes and CHO cells expressing human B1 receptor, respectively.

Formel: C₄₂H₅₂N₄O₇S·HCl
Lagertemperatur: Kühlschrank
MDL: MFCD09260115
CAS-Nummer: 464930-42-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR