AZD-5069 98%

Lieferant: Apollo Scientific

Synonyme: N-(2-((2,3-difluorobenzyl)thio)-6-(((2R,3S)-3,4-dihydroxybutan-2-yl)oxy)pyrimidin-4-yl)azetidine-1-sulphonamide

BISN0063-1MG
APOSBISN0063-1MGEA 39.3 EUR
APOSBISN0063-1MG
AZD-5069 98%
AZD-5069

AZD 5069 is a selective and reversible antagonist of CXCR2 that can inhibit CXCL8 binding to CXCR2 with pIC50 of 9.1.

Formel: C₁₈H₂₂F₂N₄O₅S₂
Lagertemperatur: Gefrierschrank
MDL: MFCD19443645
CAS-Nummer: 878385-84-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR