BAY-8002 98%

Lieferant: Apollo Scientific

BISN0075-5MG
APOSBISN0075-5MGEA 75.4 EUR
APOSBISN0075-5MG
BAY-8002 98%
BAY-8002

BAY-8002 is a potent MCT Inhibitor with IC50 value of 8nM for inhibition of L-lactate uptake.

Formel: C₂₀H₁₄ClNO₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD05157149
CAS-Nummer: 724440-27-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR