PZ-2891 98%

Lieferant: Apollo Scientific

Synonyme: 6-(4-(2-(4-Isopropylphenyl)acetyl)piperazin-1-yl)pyridazine-3-carbonitrile

BISN0079-10MG
APOSBISN0079-10MGEA 112 EUR
APOSBISN0079-10MG
PZ-2891 98%
PZ-2891

PZ-2891 is a pantothenate kinase (PANK) modulator with IC50 values of 40.2nM, 0.7nM and 1.3nM for human pantothenate kinases PANK1β, PANK2, and PANK3, respectively.

Formel: C₂₀H₂₃N₅O
Molecular Weight: 349,44 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31812779
CAS-Nummer: 2170608-82-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR