Retro-2 cycl 98%

Lieferant: Apollo Scientific

Synonyme: RN 1-001, Retro-2, 2-(5-Methylthiophen-2-yl)-3-phenyl-2,3-dihydroquinazolin-4(1H)-one

BISN0083-1MG
APOSBISN0083-1MGEA 29 EUR
APOSBISN0083-1MG
Retro-2 cycl 98%
Retro-2 cycl

Retro-2 is a specific retrograde trafficking inhibitor of protein toxins.

Formel: C₁₉H₁₆N₂OS
Molecular Weight: 320,41 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28124399
CAS-Nummer: 1429192-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR