VU0155069 98%

Lieferant: Apollo Scientific

Synonyme: (S)-N-(1-(4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzo[d]imidazol-1-yl)piperidin-1-yl)propan-2-yl)-2-naphthamide

BISN0085-1MG
APOSBISN0085-1MGEA 29 EUR
APOSBISN0085-1MG
VU0155069 98%
VU0155069

VU-0155069 is a selective phospholipase D1 (PLD1) inhibitor with IC50 values of 46 and 933 nM for PLD1 and PLD2 respectively.

Formel: C₂₆H₂₇ClN₄O₂
Molecular Weight: 462,970 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD23160355
CAS-Nummer: 1130067-06-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR