DIM-C-pPhCO2Me 99%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-(di(1H-indol-3-yl)methyl)benzoate

BISN0093-1MG
APOSBISN0093-1MGEA 29 EUR
APOSBISN0093-1MG
DIM-C-pPhCO2Me 99%
DIM-C-pPhCO2Me

DIM-C-pPhCO2Me is a NR4A1 antagonist.

Formel: C₂₅H₂₀N₂O₂
Molecular Weight: 380,45 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD31697720
CAS-Nummer: 151358-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR