Delamanid

Lieferant: Apollo Scientific

Synonyme: (R)-2-Methyl-6-nitro-2-((4-(4-(4-(trifluoromethoxy)phenoxy)piperidin-1-yl)phenoxy)methyl)-2,3-dihydroimidazo[2,1-b]oxazole

BISN0100-5MG
APOSBISN0100-5MGEA 58.4 EUR
APOSBISN0100-5MG
Delamanid
Delamanid

Delamanid a mycobacterial cell wall synthesis inhibitor inhibits the synthesisi of mucolic acids cruciala component of the cell wall of the Mycobacterium tuberculosis complex.

Formel: C₂₅H₂₅F₃N₄O₆
Molecular Weight: 534.492 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18251539
CAS-Nummer: 681492-22-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR