Pravastatin sodium

Lieferant: Apollo Scientific

BISN0103-100MG
APOSBISN0103-100MGEA 55.8 EUR
APOSBISN0103-100MG
Pravastatin sodium
Pravastatin sodium

Pravastatin Sodium is a competitive HMG-CoA reductase inhibitor with IC50 value of 56 M.

Formel: C₂₃H₃₅NaO₇
Lagertemperatur: Raumtemperatur
MDL: MFCD00887601
CAS-Nummer: 81131-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR