Chloroquin-diphosphat (Salz)

Lieferant: Apollo Scientific

Synonyme: N4-(7-Chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine bis(phosphate), N4-(7-Chloro-4-quinolyl)-N1,N1-diethyl-1,4-pentanediamine Diphosphate, 7-Chloro-4-[4-(diethylamino)-1-methylbutylamino]quinoline Diphosphate, Chloroquine phosphate

BISN0106-25MG BISN0106-100MG
APOSBISN0106-25MGEA 79.9 EUR
APOSBISN0106-25MG APOSBISN0106-100MG
Chloroquin-diphosphat (Salz)
Chloroquine diphosphate salt

Chloroquine Diphosphate is an inhibitor of autophagylysosome and Toll-like receptor TLR signalling also acting as an ATM acitivator its an antimalarial drug.

Formel: C₁₈H₂₆ClN₃
Molecular Weight: 515.86 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00069852
CAS-Nummer: 50-63-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR