Sivelestat sodium

Lieferant: Apollo Scientific

BISN0123-5MG BISN0123-10MG BISN0123-50MG
APOSBISN0123-5MGEA 109 EUR
APOSBISN0123-5MG APOSBISN0123-10MG APOSBISN0123-50MG
Sivelestat sodium
Sivelestat sodium

Sivelestat sodium is a competitive inhibitor of human neutrophil elastase (IC50 44 nM Ki200 nM) also inhibits leukocyte elastase obtained from rabbit, rat, hamster and mouse.

Formel: C₂₀H₂₁N₂NaO₇S·4H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD01937969
CAS-Nummer: 201677-61-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR