GSK126
Lieferant: Apollo Scientific
Synonyme:
(S)-1-(sec-Butyl)-N-((4,6-dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-3-methyl-6-(6-(piperazin-1-yl)pyridin-3-yl)-1H-indole-4-carboxamide
BISN0124-5MG
APOSBISN0124-5MGEA
0
EUR
APOSBISN0124-5MG
GSK126
GSK126
GSK126 is a potent, highly selective EZH2 methyltransferase inhibitor with IC50 of 9.9 nM, > 1000-fold selective for EZH2 over 20 other human methyltransferases.
Formel:
C₃₁H₃₈N₆O₂ Molecular Weight: 526,67 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23381067 CAS-Nummer: 1346574-57-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...