Fluvastatin sodium salt

Lieferant: Apollo Scientific

BISN0143-100MG
APOSBISN0143-100MGEA 25.8 EUR
APOSBISN0143-100MG
Fluvastatin sodium salt
Fluvastatin sodium salt

Fluvastatin Sodium is a synthetic indole-heptanoic-acid-derived inhibitor of hepatic HMG-CoA reductase with IC50 of 8 nM.

Formel: C₂₄H₂₅FNNaO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00929076
CAS-Nummer: 93957-55-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR