β-Lapachone
Lieferant: Apollo Scientific
Synonyme:
2,2-Dimethyl-3,4-dihydro-2H-benzo[h]chromene-5,6-dione
BISN0149-5MG
APOSBISN0149-5MGEA
79.9
EUR
APOSBISN0149-5MG
β-Lapachone
beta-Lapachone
Lapachone is a naturally occurring quinone obtained from the bark of the lapacho tree (Tabebuia avellanedae) with cancer chemopreventive properties and a potent inhibitor of IDO1 (IC500.44 M).
Formel:
C₁₅H₁₄O₃ Molecular Weight: 242,274 g/mol Schmelzpunkt: 151…153 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD01712233 CAS-Nummer: 4707-32-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...