GW 4869 dihydrocloride

Lieferant: Apollo Scientific

Synonyme: GW4869, p-Benzenediacrylanilide, 4′,4′′-di-2-imidazolin-2-yl-, dihydrochloride

BISN0156-10MG
APOSBISN0156-10MGEA 207 EUR
APOSBISN0156-10MG
GW 4869 dihydrocloride
GW 4869 dihydrocloride

GW4869 2HCl is a cell permeable, selective and non-competitve inhibitor of N-SMase (neutral sphingomyelinase), which is important in the metabolism of sphingomyelin producing ceramide and phosphocholine and in cell signaling and regulation.

Formel: C₃₀H₃₀Cl₂N₆O₂
Molecular Weight: 577.5000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06411564
CAS-Nummer: 6823-69-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR