OTS-964
Lieferant: Apollo Scientific
Synonyme:
(R)-9-(4-(1-(Dimethylamino)propan-2-yl)phenyl)-8-hydroxy-6-methylthieno[2,3-c]quinolin-4(5H)-one hydrochloride, (R)-9-(4-(1-(dimethylamino)propan-2-yl)phenyl)-8-hydroxythieno[2,3-c]quinolin-4(5H)-one, OTS964 HCl
BISN0160-2MG
APOSBISN0160-2MGEA
77
EUR
APOSBISN0160-2MG
OTS-964
OTS-964
MAPK/ERK Signalling Pathway
Formel:
C₂₂H₂₂N₂O₂S Lagertemperatur: Gefrierschrank |
MDL:
MFCD28386220 CAS-Nummer: 1338545-07-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...