NQDI-1

Lieferant: Apollo Scientific

Synonyme: Ethyl 2,7-dioxo-2,7-dihydro-3H-naphtho[1,2,3-de]quinoline-1-carboxylate

BISN0166-1MG
APOSBISN0166-1MGEA 29 EUR
APOSBISN0166-1MG
NQDI-1
NQDI-1

MAPK/ERK Signalling Pathway

Formel: C₁₉H₁₃NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD03265970
CAS-Nummer: 175026-96-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR