Bentamapimod

Lieferant: Apollo Scientific

Synonyme: AS-602801, 2-(Benzo[d]thiazol-2-yl)-2-(2-((4-(morpholinomethyl)benzyl)oxy)pyrimidin-4-yl)acetonitrile

BISN0167-1MG
APOSBISN0167-1MGEA 102 EUR
APOSBISN0167-1MG
Bentamapimod
Bentamapimod

MAPK/ERK Signalling Pathway

Formel: C₂₅H₂₃N₅O₂S
Molecular Weight: 457.6 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08272350
CAS-Nummer: 848344-36-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR