BI-78D3

Lieferant: Apollo Scientific

BISN0178-1MG
APOSBISN0178-1MGEA 36.4 EUR
APOSBISN0178-1MG
BI-78D3
BI-78D3

MAPK/ERK Signalling Pathway

Formel: C₁₃H₉N₅O₅S₂
Lagertemperatur: Gefrierschrank
MDL: MFCD01313575
CAS-Nummer: 883065-90-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR