VRT752271

Lieferant: Apollo Scientific

Synonyme: Ulixertinib, BVD 523, N-[(1S)-1-(3-chlorophenyl)-2-hydroxyethyl]-4-[5-chloro-2-(propan-2-ylamino)pyridin-4-yl]-1H-pyrrole-2-carboxamide, VRT-752271, VRT752271, BVD-523, (S)-4-(5-Chloro-2-(isopropylamino)pyridin-4-yl)-N-(1-(3-chlorophenyl)-2-hydroxyethyl)-1H-pyrrole-2-carboxamide, BVD523

BISN0179-5MG
APOSBISN0179-5MGEA 102 EUR
APOSBISN0179-5MG
VRT752271
VRT752271

MAPK/ERK Signalling Pathway

Formel: C₂₁H₂₂Cl₂N₄O₂
Molecular Weight: 433,33 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22628898
CAS-Nummer: 869886-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR