BI-D1870

Lieferant: Apollo Scientific

Synonyme: 2-((3,5-Difluoro-4-hydroxyphenyl)amino)-8-isopentyl-5,7-dimethyl-7,8-dihydropteridin-6(5H)-one

BISN0185-5MG
APOSBISN0185-5MGEA 108 EUR
APOSBISN0185-5MG
BI-D1870
BI-D1870

MAPK/ERK Signalling Pathway

Formel: C₁₉H₂₃F₂N₅O₂
Molecular Weight: 391,423 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD11223662
CAS-Nummer: 501437-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR