JNK-IN-7
Lieferant: Apollo Scientific
Synonyme:
3-[[(E)-4-(dimethylamino)but-2-enoyl]amino]-N-[4-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]benzamide, JNKIN-7, JNK-IN-7, JNK-IN7, JNK IN 7, JNKIN7, 3-(4-(Dimethylamino)but-2-enamido)-N-(4-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)benzamide
BISN0187-2MG
APOSBISN0187-2MGEA
156
EUR
APOSBISN0187-2MG
JNK-IN-7
JNK-IN-7
MAPK/ERK Signalling Pathway
Formel:
C₂₈H₂₇N₇O₂ Molecular Weight: 493,56 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD25976765 CAS-Nummer: 1408064-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...