CC-401 hydrochloride

Lieferant: Apollo Scientific

Synonyme: 3-(3-(2-(Piperidin-1-yl)ethoxy)phenyl)-5-(1H-1,2,4-triazol-5-yl)-1H-indazole hydrochloride, CC-401 HCl, CC401

BISN0198-2MG
APOSBISN0198-2MGEA 81.2 EUR
APOSBISN0198-2MG
CC-401 hydrochloride
CC-401 hydrochloride

MAPK/ERK Signalling Pathway

Formel: C₂₂H₂₄N₆O·HCl
Molecular Weight: 424,9 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD21364765
CAS-Nummer: 1438391-30-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR