APS-2-79 hydrochloride

Lieferant: Apollo Scientific

Synonyme: APS-2-79, 6,7-dimethoxy-N-(2-methyl-4-phenoxyphenyl)quinazolin-4-aminehydrochloride, 6,7-Dimethoxy-N-(2-methyl-4-phenoxyphenyl)quinazolin-4-amine hydrochloride, 6,7-dimethoxy-N-(2-methyl-4-phenoxyphenyl)quinazolin-4-amine hydrochloride

BISN0199-5MG
APOSBISN0199-5MGEA 34.8 EUR
APOSBISN0199-5MG
APS-2-79 hydrochloride
APS-2-79 hydrochloride

MAPK/ERK Signalling Pathway

Formel: C₂₃H₂₂ClN₃O₃
Molecular Weight: 423.89 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30536203
CAS-Nummer: 2002381-31-7
UN: 3077
ADR: 9+EHS III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR