(R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-(2-fluoro-4-iodophenylamino)benzamide
Lieferant: Apollo Scientific
Synonyme:
(R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-((2-fluoro-4-iodophenyl)amino)benzamide, Mirdametinib, PD-0325901
BISN0215-5MG
BISN0215-10MG
BISN0215-25MG
APOSBISN0215-5MGEA
161
EUR
APOSBISN0215-5MG
APOSBISN0215-10MG
APOSBISN0215-25MG
(R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-(2-fluoro-4-iodophenylamino)benzamide
(R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-(2-fluoro-4-iodophenylamino)benzamide
MAPK/ERK Signalling Pathway
Formel:
C₁₆H₁₄F₃IN₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08435926 CAS-Nummer: 391210-10-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...