AZ-304

Lieferant: Apollo Scientific

BISN0220-50MG
APOSBISN0220-50MGEA 132 EUR
APOSBISN0220-50MG
AZ-304
AZ-304

MAPK/ERK Signalling Pathway

Formel: C₂₇H₂₅N₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31812677
CAS-Nummer: 942507-42-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR