XMD17-109

Lieferant: Apollo Scientific

Synonyme: XMD17109, XMD17-109, 11-cyclopentyl-2-[2-ethoxy-4-[4-(4-methylpiperazin-1-yl)piperidine-1-carbonyl]anilino]-5-methylpyrimido[4,5-b][1,4]benzodiazepin-6-one, XMD17 109, 11-Cyclopentyl-2-((2-ethoxy-4-(4-(4-methylpiperazin-1-yl)piperidine-1-carbonyl)phenyl)amino)-5-methyl-5,11-dihydro-6H-benzo[e]pyrimido[5,4-b][1,4]diazepin-6-one

BISN0221-10MG
APOSBISN0221-10MGEA 154 EUR
APOSBISN0221-10MG
XMD17-109
XMD17-109

MAPK/ERK Signalling Pathway

Formel: C₃₆H₄₆N₈O₃
Molecular Weight: 638.8 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26142929
CAS-Nummer: 1435488-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR