SB239063

Lieferant: Apollo Scientific

Synonyme: 4-[4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)imidazol-1-yl]cyclohexan-1-ol, trans-4-[4-(4-fluorophenyl)-5-(2-methoxy-4-pyrimidinyl)-1H-imidazol-1-yl]-cyclohexanol, Rel-(1r,4r)-4-(4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)-1H-imidazol-1-yl)cyclohexan-1-ol, 4-(4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)-1H-imidazol-1-yl)cyclohexanol

BISN0226-5MG
APOSBISN0226-5MGEA 81.2 EUR
APOSBISN0226-5MG
SB239063
SB239063

MAPK/ERK Signalling Pathway

Formel: C₂₀H₂₁FN₄O₂
Molecular Weight: 368.4 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03792786
CAS-Nummer: 193551-21-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR