URMC-099

Lieferant: Apollo Scientific

Synonyme: 3-(1H-Indol-5-yl)-5-(4-(4-methylpiperazin-1-yl)methyl)phenyl)-1H-pyrrolo[2,3- b]pyridine, 3-(1H-Indol-5-yl)-5-(4-((4-methylpiperazin-1-yl)methyl)phenyl)-1H-pyrrolo[2,3-b]pyridine

BISN0232-100MG
APOSBISN0232-100MGEA 287 EUR
APOSBISN0232-100MG
URMC-099
URMC-099

MAPK/ERK Signalling Pathway

Formel: C₂₇H₂₇N₅
Molecular Weight: 421.5 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28168077
CAS-Nummer: 1229582-33-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR