GDC0879

Lieferant: Apollo Scientific

Synonyme: 2-[4-[(1E)-1-hydroxyimino-2,3-dihydroinden-5-yl]-3-pyridin-4-ylpyrazol-1-yl]ethanol, GDC-0879, 5-(1-(2-Hydroxyethyl)-3-(pyridin-4-yl)-1H-pyrazol-4-yl)-2,3-dihydro-1H-inden-1-one oxime

BISN0238-10MG
APOSBISN0238-10MGEA 426 EUR
APOSBISN0238-10MG
GDC0879
GDC0879

MAPK/ERK Signalling Pathway

Formel: C₁₉H₁₈N₄O₂
Molecular Weight: 334.37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16875916
CAS-Nummer: 905281-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR