AZ 628

Lieferant: Apollo Scientific

Synonyme: AZ 628, 3-(2-cyanopropan-2-yl)-N-[4-methyl-3-[(3-methyl-4-oxoquinazolin-6-yl)amino]phenyl]benzamide, AZ-628, AZ628, 3-(2-Cyanopropan-2-yl)-N-(4-methyl-3-((3-methyl-4-oxo-3,4-dihydroquinazolin-6-yl)amino)phenyl)benzamide

BISN0260-25MG
APOSBISN0260-25MGEA 148 EUR
APOSBISN0260-25MG
AZ 628
AZ 628

MAPK/ERK Signalling Pathway

Formel: C₂₇H₂₅N₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD17392577
CAS-Nummer: 878739-06-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR