IQ-1S
Lieferant: Apollo Scientific
Synonyme:
11H-Indeno[1,2-b]quinoxalin-11-one oxime sodium
BISN0275-10MG
APOSBISN0275-10MGEA
226
EUR
APOSBISN0275-10MG
IQ-1S
IQ-1S
MAPK/ERK Signalling Pathway
Formel:
C₁₅H₈N₃NaO Molecular Weight: 269.23 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
1421610-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...