SB242235

Lieferant: Apollo Scientific

Synonyme: 4-(4-(4-Fluorophenyl)-1-(piperidin-4-yl)-1H-imidazol-5-yl)-2-methoxypyrimidine, SB 242235, 4-[5-(4-fluorophenyl)-3-piperidin-4-ylimidazol-4-yl]-2-methoxypyrimidine

BISN0286-1MG BISN0286-5MG BISN0286-25MG
APOSBISN0286-1MGEA 87 EUR
APOSBISN0286-1MG APOSBISN0286-5MG APOSBISN0286-25MG
SB242235
SB242235

MAPK/ERK Signalling Pathway

Formel: C₁₉H₂₀FN₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD08690575
CAS-Nummer: 193746-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR