Kämpferol
Lieferant: Apollo Scientific
Synonyme:
3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
BISN0318-100MG
BISN0318-250MG
APOSBISN0318-100MGEA
109
EUR
APOSBISN0318-100MG
APOSBISN0318-250MG
Kämpferol
Kämpferol
Formel:
C₁₅H₁₀O₆ Molecular Weight: 286.24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016938 CAS-Nummer: 520-18-3 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...