Thiamin Hydrochlorid ≥99%

Lieferant: Apollo Scientific
Achtung

BIT0614-100G BIT0614-1KG BIT0614-250G
APOSBIT0614-100GEA 0 EUR
APOSBIT0614-100G APOSBIT0614-1KG APOSBIT0614-250G
Thiamin Hydrochlorid ≥99%
Thiamin Hydrochlorid
Formel: C₁₂H₁₈Cl₂N₄OS
Lagertemperatur: Raumtemperatur
MDL: MFCD00012780
CAS-Nummer: 67-03-8
EINECS: 200-641-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR