Thiamin Hydrochlorid ≥99%
Lieferant: Apollo Scientific
|
Achtung
|
BIT0614-100G
BIT0614-1KG
BIT0614-250G
APOSBIT0614-100GEA
0
EUR
APOSBIT0614-100G
APOSBIT0614-1KG
APOSBIT0614-250G
Thiamin Hydrochlorid ≥99%
Thiamin Hydrochlorid
Formel:
C₁₂H₁₈Cl₂N₄OS Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012780 CAS-Nummer: 67-03-8 EINECS: 200-641-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...