L(-)-Tryptophan ≥99,5%

Lieferant: Apollo Scientific
Achtung

BIT0720-100G BIT0720-1KG BIT0720-500G
APOSBIT0720-100GEA 0 EUR
APOSBIT0720-100G APOSBIT0720-1KG APOSBIT0720-500G
L(-)-Tryptophan ≥99,5%
L(-)-Tryptophan

USP, pyrogen free

Formel: C₁₁H₁₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00064340
CAS-Nummer: 73-22-3
EINECS: 200-795-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR