D(+)-Tyrosin ≥99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(R)-(+)-p-Hydroxyphenylalanin, H-D-Tyr-OH, (R)-(+)-2-Amino-3-(4-hydroxyphenyl)propionsäure, D(+)-p-Hydroxyphenylalanin
BIT0722-25G
APOSBIT0722-25GEA
0
EUR
APOSBIT0722-25G
D(+)-Tyrosin ≥99%
D(+)-Tyrosin
Formel:
C₉H₁₁NO₃ Molecular Weight: 181,19 g/mol Schmelzpunkt: 310…314 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063073 CAS-Nummer: 556-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...