DL-Tyrosin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydroxyphenylalanin, 2-Amino-3-(4-hydroxyphenyl)propionsäure, H-DL-Tyr-OH, 3-(p-Hydroxyphenyl)-DL-alanin
BIT0723-1KG
APOSBIT0723-1KGEA
480
EUR
APOSBIT0723-1KG
DL-Tyrosin
DL-Tyrosin
Formel:
C₉H₁₁NO₃ Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063074 CAS-Nummer: 556-03-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...