Tween® 20 (Polysorbat)
Lieferant: Apollo Scientific
BIT0803-1L
APOSBIT0803-1LEA
50.9
EUR
APOSBIT0803-1L
Tween® 20 (Polysorbat)
Tween® 20 (Polysorbat)
Non ionic detergent used for cell lysis, nuclei isolationand cell fractionation.
Formel:
C₅₈H₁₁₄O₂₆ Siedepunkt: >100 °C Schmelzpunkt: –14 °C Dichte: 1,095 g/cm³ (25 °C) Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00165986 CAS-Nummer: 9005-64-5 EINECS: 500-018-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...