1,3,5-Triphenylformazan

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3,5-Triphenyl tetrazolium formazan

BIT3087-1G
APOSBIT3087-1GEA 49.3 EUR
APOSBIT3087-1G
1,3,5-Triphenylformazan
1,3,5-Triphenylformazan
Formel: C₁₉H₁₆N₄
Molecular Weight: 300,37 g/mol
Schmelzpunkt: 163…166 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00020647
CAS-Nummer: 531-52-2
UN: 3077
ADR: 9+EHS,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR