2,4,6-Tris(2-pyridyl)-1,3,5-triazin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,4,6-Tris(2-pyridyl)-s-triazin
BIT3089-10G
BIT3089-25G
BIT3089-5G
APOSBIT3089-10GEA
43.5
EUR
APOSBIT3089-10G
APOSBIT3089-25G
APOSBIT3089-5G
2,4,6-Tris(2-pyridyl)-1,3,5-triazin
2,4,6-Tris(2-pyridyl)-1,3,5-triazin
Formel:
C₁₈H₁₂N₆ Molecular Weight: 312.33 g/mol Schmelzpunkt: 247…249 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00006045 CAS-Nummer: 3682-35-7 EINECS: 222-965-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...